Allyl benzoate ≥98.0%

Leverantör: TCI

Synonyms: Benzoic acid Allyl Ester, Benzoic Acid Allyl Ester, Allyl Benzoate

B2041-25G B2041-250G
TCIAB2041-25GEA 695 SEK
TCIAB2041-25G TCIAB2041-250G
Allyl benzoate ≥98.0%
Allyl benzoate
Formel: C₁₀H₁₀O₂
Kokpunkt: 242°C
MDL Number: MFCD00026104
CAS nummer: 583-04-0
EINECS: 209-494-4

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR