4-Bromophthalimide ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI
|
Varning
|
Synonyms:
5-Bromoisoindoline-1,3-dione, 5-bromo-2,3-dihydro-1H-isoindole-1,3-dione
B2009.0005
B2009-25G
B2009-5G
TCIAB2009.0005EA
0
SEK
TCIAB2009.0005
TCIAB2009-25G
TCIAB2009-5G
4-Bromophthalimide ≥98.0% (by HPLC, titration analysis)
4-Bromophthalimide
Formel:
C₈H₄BrNO₂ MW: 226.03 g/mol |
MDL Number:
MFCD00466049 CAS nummer: 6941-75-9 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 232.0 to 238.0 deg-C |
Solubility in N,N-DMF | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...