Ethyl-2-benzylacetoacetate ≥97.0%
Leverantör: TCI
Synonyms:
Ethyl 2-benzyl-3-oxobutanoate, 2-Benzylacetoacetic Acid Ethyl Ester, Ethyl 2-Acetyl-3-phenylpropionate, 2-Acetyl-3-phenylpropionic Acid Ethyl Ester
B2008-25G
B2008-5G
TCIAB2008-25GEA
2050
SEK
TCIAB2008-25G
TCIAB2008-5G
Ethyl-2-benzylacetoacetate ≥97.0%
Ethyl-2-benzylacetoacetate
Formel:
C₁₃H₁₆O₃ Kokpunkt: 284°C |
MDL Number:
MFCD00009156 CAS nummer: 620-79-1 EINECS: 210-651-4 |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...