4-(2-Bromoethyl)benzenesulphonic acid ≥97.0%

Leverantör: TCI EUROPE
Fara

B2006-1G B2006-5G
TCIAB2006-1GEA 2225 SEK
TCIAB2006-1G TCIAB2006-5G
4-(2-Bromoethyl)benzenesulphonic acid ≥97.0%
4-(2-Bromoethyl)benzenesulphonic acid
Formel: C₈H₉BrO₃S
MW: 265,12 g/mol
MDL Number: MFCD01861279
CAS nummer: 54322-31-5
EINECS: 259-094-9
UN: 2585
ADR: 8,III

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) min. 97.0 % (calcd.on anh.substance)
Melting point 82.0 to 86.0 deg-C
Water max. 8.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR