4-Butyrylbiphenyl ≥99.0%

Leverantör: TCI EUROPE

Synonyms: 4'-Phenylbutyrophenone

B1956-25G B1956-500G
TCIAB1956-25GEA 430 SEK
TCIAB1956-25G TCIAB1956-500G
4-Butyrylbiphenyl ≥99.0%
4-Butyrylbiphenyl
Formel: C₁₆H₁₆O
MW: 224,30 g/mol
Kokpunkt: 355°C
MDL Number: MFCD00022005
CAS nummer: 13211-01-3
EINECS: 236-178-3

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 99.0 %
Melting point 93.0 to 96.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR