Benzeneruthenium(II) chloride dimer ≥95.0%

Leverantör: TCI
Varning

Synonyms: Dichloro(benzene)ruthenium(II) dimer

B1902-1G B1902-5G
TCIAB1902-1GEA 2050 SEK
TCIAB1902-1G TCIAB1902-5G
Benzeneruthenium(II) chloride dimer ≥95.0%
Benzeneruthenium(II) chloride dimer
Formel: C₁₂H₁₂Cl₄Ru₂
MDL Number: MFCD00064686
CAS nummer: 37366-09-9

Order Now

Specifikation testresultat

Appearance Orange to Brown to Dark red powder to crystal
Purity (Argentmetric Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR