5-Bromo-2,2'-bithiophene-5'-carboxaldehyde ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 5'-Bromo-[2,2'-bithiophene]-5-carbaldehyde, 5-Bromo-5'-formyl-2,2'-bithiophene

B1874-1G B1874-5G
TCIAB1874-1GEA 790 SEK
TCIAB1874-1G TCIAB1874-5G
5-Bromo-2,2'-bithiophene-5'-carboxaldehyde ≥98.0%
5-Bromo-2,2'-bithiophene-5'-carboxaldehyde
Formel: C₉H₅BrOS₂
MW: 273,16 g/mol
MDL Number: MFCD01321134
CAS nummer: 110046-60-1

Order Now

Specifikation testresultat

Appearance Green to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 145.0 to 149.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR