Methyl decanoate ≥99%
Leverantör: Thermo Fisher Scientific
A15658.18
A15658.22
A15658.30
A15658.18EA
1075
SEK
A15658.18
A15658.22
A15658.30
Methyl decanoate ≥99%
Methyl decanoate
Purity: 99%
CAS No.: 110-42-9
Molecular Formula: CH3(CH2)8CO2CH3
Formula Weight: 186.30
Flash Point: 94°(201°F)
Refractive Index: 1.4250
Melting Point: -18°
Boiling Point: 223-225°
Beilstein Reference: 1759170
MDL No.: MFCD00009580
Formel:
C₁₁H₂₂O₂ Kokpunkt: 223…225 °C Smältpunkt: –18 °C Densitet: 0,877 Flampunkt: 94 °C (201 °F) Storage Temperature: Omgivande |
MDL Number:
MFCD00009580 CAS nummer: 110-42-9 EINECS: 203-766-6 |