4-tert-Butylbiphenyl ≥98.0%
Leverantör: TCI EUROPE
B1811-25G
B1811-5G
TCIAB1811-25GEA
3100
SEK
TCIAB1811-25G
TCIAB1811-5G
4-tert-Butylbiphenyl ≥98.0%
4-tert-Butylbiphenyl
Formel:
C₁₆H₁₈ MW: 210,32 g/mol Kokpunkt: 310°C |
MDL Number:
MFCD00222366 CAS nummer: 1625-92-9 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 50.0 to 54.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...