4-Bromomethyl-2-cyanobiphenyl ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 4'-(Bromomethyl)-[1,1'-biphenyl]-2-carbonitrile

B1806-25G B1806-5G
TCIAB1806-25GEA 655 SEK
TCIAB1806-25G TCIAB1806-5G
4-Bromomethyl-2-cyanobiphenyl ≥98.0%
4-Bromomethyl-2-cyanobiphenyl
Formel: C₁₄H₁₀BrN
MW: 272,14 g/mol
MDL Number: MFCD00671503
CAS nummer: 114772-54-2
EINECS: 425-280-5

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 125.0 to 128.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR