(1-Bromovinyl)trimethylsilane ≥96.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
1-Bromo-1-trimethylsilylethylene, alpha-Trimethylsilylvinyl Bromide
B1769-1G
B1769-5G
TCIAB1769-1GEA
445
SEK
TCIAB1769-1G
TCIAB1769-5G
(1-Bromovinyl)trimethylsilane ≥96.0%
(1-Bromovinyl)trimethylsilane
Formel:
C₅H₁₁BrSi MW: 179,13 g/mol Kokpunkt: 128°C Flampunkt: 9°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00014927 CAS nummer: 13683-41-5 EINECS: 237-195-9 UN: 1993 ADR: 3,II |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...