1,2-Bis(trimethylsilyloxy)cyclobutene ≥94.0%
Leverantör: TCI
|
Varning
|
Synonyms:
1,2-Bis((trimethylsilyl)oxy)cyclobut-1-ene
B1768-1G
B1768-5G
TCIAB1768-1GEA
550
SEK
TCIAB1768-1G
TCIAB1768-5G
1,2-Bis(trimethylsilyloxy)cyclobutene ≥94.0%
1,2-Bis(trimethylsilyloxy)cyclobutene
Formel:
C₁₀H₂₂O₂Si₂ Flampunkt: 61°C |
MDL Number:
MFCD00010798 CAS nummer: 17082-61-0 |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 94.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...