(S)-(-)-3-(tert-Butoxycarbonyl)-4-formyl-2,2-dimethyl-1,3-oxazolidine ≥95.0%

Leverantör: TCI
Varning

Synonyms: (S)-(-)-3-Boc-4-formyl-2,2-dimethyl-1,3-oxazolidine, (S)-(-)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde, tert-Butyl (S)-(-)-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, (S)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde

B1759-1G B1759-5G
TCIAB1759-1GEA 1025 SEK
TCIAB1759-1G TCIAB1759-5G
(S)-(-)-3-(tert-Butoxycarbonyl)-4-formyl-2,2-dimethyl-1,3-oxazolidine ≥95.0%
(S)-(-)-3-(tert-Butoxycarbonyl)-4-formyl-2,2-dimethyl-1,3-oxazolidine
Formel: C₁₁H₁₉NO₄
Kokpunkt: 97°C
Flampunkt: 107°C
Storage Temperature: Kylskåp
MDL Number: MFCD00209557
CAS nummer: 102308-32-7

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %
Specific rotation [a]20/D -85.0 to -98.0 deg (C=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR