Bis(4-diethylaminophenyl)methanol ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 4,4'-Bis(diethylamino)benzhydrol
B1725-1G B1725-5G
TCIAB1725-1GEA 1050 SEK
TCIAB1725-1G TCIAB1725-5G
Bis(4-diethylaminophenyl)methanol ≥98.0%
Bis(4-diethylaminophenyl)methanol
Formel: C₂₁H₃₀N₂O
MW: 326,48 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00035979
CAS nummer: 134-91-8
EINECS: 205-162-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ee52fc2eae51b24bc834b27d51344ae8a5911c7&vid=5ee52fc2eae51b24bc834b27d51344ae8a5911c7&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 78.0 to 83.0 deg-C