Sodium-3-bromopropanesulphonate ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 3-Bromopropanesulphonic acid Sodium Salt

B1707-25G B1707-100G
TCIAB1707-25GEA 925 SEK
TCIAB1707-25G TCIAB1707-100G
Sodium-3-bromopropanesulphonate ≥98.0%
Sodium-3-bromopropanesulphonate
Formel: C₃H₇BrO₃S
MW: 225,03 g/mol
MDL Number: MFCD00024999
CAS nummer: 55788-44-8

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (Ion exchange titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR