1-(3-Bromopropyl)pyrrole ≥97.0%
Leverantör: TCI EUROPE
Varning
B1623-1G B1623-5G
TCIAB1623-1GEA 2025 SEK
TCIAB1623-1G TCIAB1623-5G
1-(3-Bromopropyl)pyrrole ≥97.0%
1-(3-Bromopropyl)pyrrole
Formel: C₇H₁₀BrN
MW: 188,07 g/mol
Kokpunkt: 78°C
Storage Temperature: Kylskåp
MDL Number: MFCD00191324
CAS nummer: 100779-91-7
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %