4,4'-Bicyclohexanol mixture of isomers ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1,1'-Bicyclohexyl-4,4'-diol, [1,1'-Bi(cyclohexane)]-4,4'-diol, 4,4'-Dihydroxydicyclohexyl
B1621-25G
B1621-500G
TCIAB1621-25GEA
645
SEK
TCIAB1621-25G
TCIAB1621-500G
4,4'-Bicyclohexanol mixture of isomers ≥98.0%
4,4'-Bicyclohexanol
Formel:
C₁₂H₂₂O₂ MW: 198,31 g/mol |
MDL Number:
MFCD00012056 CAS nummer: 20601-38-1 EINECS: 243-908-4 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % (total of isomer) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...