2-Buten-1-ylsuccinic anhydride ≥98.0%

Leverantör: TCI
Varning

Synonyms: 4-Hexene-1,2-dicarboxylic Anhydride

B1613-25G B1613-5G
TCIAB1613-25GEA 2475 SEK
TCIAB1613-25G TCIAB1613-5G
2-Buten-1-ylsuccinic anhydride ≥98.0%
2-Buten-1-ylsuccinic anhydride
Formel: C₈H₁₀O₃
MDL Number: MFCD03423563
CAS nummer: 7538-42-3

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (Morpholine Method) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR