1-Benzyl-3-pyrrolidinol ≥96.0%
Leverantör: TCI
Varning
B1597-25G B1597-5G
TCIAB1597-25GEA 765 SEK
TCIAB1597-25G TCIAB1597-5G
1-Benzyl-3-pyrrolidinol ≥96.0%
1-Benzyl-3-pyrrolidinol
Formel: C₁₁H₁₅NO
Kokpunkt: 143°C
MDL Number: MFCD00012132
CAS nummer: 775-15-5
EINECS: 212-273-5
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %