tert-Butylmalononitrile 95,0 GC_ANALYSMETOD

Leverantör: TCI
Fara

B1584-25G B1584-5G
TCIAB1584-25GEA 3800 SEK
TCIAB1584-25G TCIAB1584-5G
tert-Butylmalononitrile 95,0 GC_ANALYSMETOD
tert-Butylmalononitrile
Formel: C₇H₁₀N₂
MW: 122.17 g/mol
MDL Number: MFCD00191326
CAS nummer: 4210-60-0
UN: 3439
ADR: 6.1 III

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystaline
Purity (GC) min. 95.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR