(3S)-(+)-1-Benzyl-3-(ethylamino)pyrrolidine ≥95.0%
Leverantör: TCI EUROPE
Fara
B1581-5G B1581-1G
TCIAB1581-5GEA 11075 SEK
TCIAB1581-5G TCIAB1581-1G
(3S)-(+)-1-Benzyl-3-(ethylamino)pyrrolidine ≥95.0%
(3S)-(+)-1-Benzyl-3-(ethylamino)pyrrolidine
Formel: C₁₃H₂₀N₂
MW: 204,32 g/mol
MDL Number: MFCD00191307
CAS nummer: 169750-99-6
UN: 2735
ADR: 8,III
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %
Specific rotation [a]20/D +7.0 to +9.0 deg (C=10, EtOH)