1-Benzyl-3-(methylamino)pyrrolidine ≥96.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 1-Benzyl-N-methylpyrrolidin-3-amine

B1558-25G B1558-5G
TCIAB1558-25GEA 3750 SEK
TCIAB1558-25G TCIAB1558-5G
1-Benzyl-3-(methylamino)pyrrolidine ≥96.0%
1-Benzyl-3-(methylamino)pyrrolidine
Formel: C₁₂H₁₈N₂
MW: 190,29 g/mol
Kokpunkt: 107°C
MDL Number: MFCD00191308
CAS nummer: 96568-35-3
UN: 2735
ADR: 8,III

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR