Methyl-3-benzoylpropionate ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Methyl 4-Oxo-4-phenylbutyrate, 3-Benzoylpropionic Acid Methyl Ester, Methyl 4-oxo-4-phenylbutanoate, 4-Oxo-4-phenylbutyric Acid Methyl Ester

B1546-25G B1546-5G
TCIAB1546-25GEA 2675 SEK
TCIAB1546-25G TCIAB1546-5G
Methyl-3-benzoylpropionate ≥98.0%
Methyl-3-benzoylpropionate
Formel: C₁₁H₁₂O₃
MW: 192,21 g/mol
MDL Number: MFCD00014906
CAS nummer: 25333-24-8

Order Now

Specifikation testresultat

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR