N,N'-Bis(2-aminoethyl)-1,3-propanediamine ≥97.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 3,7-Diaza-1,9-nonanediamine, N1,N1'-(Propane-1,3-diyl)bis(ethane-1,2-diamine), 1,4,8,11-Tetraazaundecane

B1445-5G B1445-1G
TCIAB1445-5GEA 3200 SEK
TCIAB1445-5G TCIAB1445-1G
N,N'-Bis(2-aminoethyl)-1,3-propanediamine ≥97.0%
N,N'-Bis(2-aminoethyl)-1,3-propanediamine
Formel: C₇H₂₀N₄
MW: 160,27 g/mol
Kokpunkt: 287°C
MDL Number: MFCD00008174
CAS nummer: 4741-99-5
EINECS: 225-254-1
UN: 2735
ADR: 8,III

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR