tert-Butoxydiphenylchlorosilane ≥95.0% stabilised

Leverantör: TCI EUROPE
Fara

Synonyms: tert-Butoxychlorodiphenylsilane, tert-Butoxydiphenylsilyl Chloride

Stabilization: Stabilised with calcium carbonate

B1436-25ML B1436-5ML
TCIAB1436-25MLEA 9200 SEK
TCIAB1436-25ML TCIAB1436-5ML
tert-Butoxydiphenylchlorosilane ≥95.0% stabilised
tert-Butoxydiphenylchlorosilane
Formel: C₁₆H₁₉ClOSi
MW: 290,86 g/mol
Kokpunkt: 112°C
Flampunkt: 56°C
Storage Temperature: Frys
MDL Number: MFCD00077672
CAS nummer: 17922-24-6
UN: 2986
ADR: 8,II

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR