Benzylhydrazine monohydrochloride ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Benzylhydrazine hydrochloride

B1396-25G B1396-5G
TCIAB1396-25GEA 4750 SEK
TCIAB1396-25G TCIAB1396-5G
Benzylhydrazine monohydrochloride ≥98.0%
Benzylhydrazine monohydrochloride
Formel: C₇H₁₀N₂
MW: 158,63 g/mol
MDL Number: MFCD00012921
CAS nummer: 1073-62-7
UN: 2811
ADR: 6.1,III

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Potassium iodate Method) min. 97.0 %
Melting point 111.0 to 115.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR