3,4-Bis(4-acetoxyphenyl)-2,4-hexadiene ≥95.0%

Leverantör: TCI

B1366-1G B1366-100MG
TCIAB1366-1GEA 3000 SEK
TCIAB1366-1G TCIAB1366-100MG
3,4-Bis(4-acetoxyphenyl)-2,4-hexadiene ≥95.0%
3,4-Bis(4-acetoxyphenyl)-2,4-hexadiene
Formel: C₂₂H₂₂O₄
MDL Number: MFCD00063286
CAS nummer: 84-19-5
EINECS: 201-520-2
Merck Index: 14,03104

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %
Melting point 119.0 to 123.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR