3-Boc-aminopyrrolidine ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 3-(BOC-amino)pyrrolidine, 2-Methyl-2-propanyl 3-pyrrolidinylcarbamate, 1,1-Dimethylethyl3-pyrrolidinylcarbamate, 3-Boc-Aminopyrrolidine

B1358-25G B1358-5G
TCIAB1358-25GEA 2400 SEK
TCIAB1358-25G TCIAB1358-5G
3-Boc-aminopyrrolidine ≥98.0% (by GC, titration analysis)
3-Boc-aminopyrrolidine
Formel: C₉H₁₈N₂O₂
MW: 186,25 g/mol
MDL Number: MFCD00059040
CAS nummer: 99724-19-3

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR