1-Benzyl-3-acetamidopyrrolidine ≥98.0%

Leverantör: TCI
Varning

B1355-25G B1355-5G
TCIAB1355-25GEA 1925 SEK
TCIAB1355-25G TCIAB1355-5G
1-Benzyl-3-acetamidopyrrolidine ≥98.0%
1-Benzyl-3-acetamidopyrrolidine
Formel: C₁₃H₁₈N₂O
MDL Number: MFCD00059037
CAS nummer: 28506-01-6

Order Now

Specifikation testresultat

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 89.0 to 92.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR