Benzofuroxan ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Benzofurazan 1-Oxide, 2,1,3-Benzoxadiazole 1-Oxide
B1316-25G
B1316-500G
TCIAB1316-25GEA
575
SEK
TCIAB1316-25G
TCIAB1316-500G
Benzofuroxan ≥98.0%
Benzofuroxan
Formel:
C₆H₄N₂O₂ MW: 136,11 g/mol |
MDL Number:
MFCD00005807 CAS nummer: 480-96-6 EINECS: 207-559-1 |
Specifikation testresultat
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 67.0 to 70.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...