1,3-Bis(diphenylphosphino)propanenickel(II) chloride ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: [1,3-Bis(diphenylphosphino)propane]nickel (II) chloride, Dichloro[bis(1,3-diphenylphosphino)propane]nickel(II), 1,3-Bis(diphenylphosphino)propane nickel(II)chloride, Dichloro[1,3-bis(diphenylphosphino)propane]nickel(II), [1,3-Bis(diphenylphosphino)propane]dichloronickel(II), [1,3-Bis(diphenylphosphino)propane]nickel(II) dichloride

B1313-25G B1313-5G
TCIAB1313-25GEA 2800 SEK
TCIAB1313-25G TCIAB1313-5G
1,3-Bis(diphenylphosphino)propanenickel(II) chloride ≥98.0%
1,3-Bis(diphenylphosphino)propanenickel(II) chloride
Formel: C₂₇H₂₆Cl₂NiP₂
MW: 542,042 g/mol
MDL Number: MFCD00015318
CAS nummer: 15629-92-2

Order Now

Specifikation testresultat

Appearance Orange to Brown to Dark red powder to crystal
Purity (Chelometric Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR