tert-Butyl-2-bromopropionate ≥97.0%
Leverantör: TCI
Varning
Synonyms: 2-Bromopropionic Acid tert-Butyl Ester, tert-Butyl 2-bromopropanoate
B1277-25G B1277-500G
TCIAB1277-25GEA 1275 SEK
TCIAB1277-25G TCIAB1277-500G
tert-Butyl-2-bromopropionate ≥97.0%
tert-Butyl-2-bromopropionate
Formel: C₇H₁₃BrO₂
Kokpunkt: 62°C
Storage Temperature: Kylskåp
MDL Number: MFCD00043897
CAS nummer: 39149-80-9
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %