7-Bromo-5-chloro-8-hydroxyquinoline ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 7-Bromo-5-chloroquinolin-8-ol

B1190-5G B1190-1G
TCIAB1190-5GEA 2300 SEK
TCIAB1190-5G TCIAB1190-1G
7-Bromo-5-chloro-8-hydroxyquinoline ≥98.0% (by GC, titration analysis)
7-Bromo-5-chloro-8-hydroxyquinoline
Formel: C₉H₅BrClNO
MW: 258,50 g/mol
MDL Number: MFCD00044601
CAS nummer: 7640-33-7
EINECS: 231-574-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=786f62b90e64f1a14b136753b410f4641557760b&vid=786f62b90e64f1a14b136753b410f4641557760b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 179.0 to 182.0 deg-C
Solubility in hot Acetic acid almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR