(+)-3-Bromocamphor-8-sulphonic acid ammonium salt ≥98.0%

Leverantör: TCI
Varning

B1125-25G B1125-5G
TCIAB1125-25GEA 1225 SEK
TCIAB1125-25G TCIAB1125-5G
(+)-3-Bromocamphor-8-sulphonic acid ammonium salt ≥98.0%
(+)-3-Bromocamphor-8-sulphonic acid ammonium salt
Formel: C₁₀H₁₈BrNO₄S
MDL Number: MFCD00167438
CAS nummer: 14575-84-9
EINECS: 238-616-9

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (Ion exchange titration) min. 98.0 %
Specific rotation [a]20/D +84.0 to +87.0 deg (C=4, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR