(+)-cis-2-Benzylaminocyclohexanemethanol ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: (+)-N-Benzyl-cis-2-hydroxymethylcyclohexylamine

B1119-1G B1119-5G
TCIAB1119-1GEA 1725 SEK
TCIAB1119-1G TCIAB1119-5G
(+)-cis-2-Benzylaminocyclohexanemethanol ≥98.0%
(+)-cis-2-Benzylaminocyclohexanemethanol
Formel: C₁₄H₂₁NO
MW: 219,33 g/mol
MDL Number: MFCD00151395
CAS nummer: 71581-92-5

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Optical purity (LC) min. 95.0 ee%
Purity (Neutralization titration) min. 98.0 %
Melting point 66.0 to 69.0 deg-C
Specific rotation [a]20/D +22.0 to +26.0 deg (C=1, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR