2-(5-Bromo-2-pyridylazo)-5-dimethylaminophenol ≥93.0% for colorimetric determination of Cd

Leverantör: TCI EUROPE

Synonyms: 2-(4-Dimethylamino-2-hydroxyphenylazo)-5-bromopyridine

B1082-1G B1082-100MG
TCIAB1082-1GEA 17400 SEK
TCIAB1082-1G TCIAB1082-100MG
2-(5-Bromo-2-pyridylazo)-5-dimethylaminophenol ≥93.0% for colorimetric determination of Cd
2-(5-Bromo-2-pyridylazo)-5-dimethylaminophenol
Formel: C₁₃H₁₃BrN₄O
MW: 321,18 g/mol
MDL Number: MFCD00059779
CAS nummer: 50783-82-9

Order Now

Specifikation testresultat

Appearance Red to Dark red to Dark purple powder to crystal
Purity (HPLC) min. 93.0 area%
Melting point 216.0 to 220.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR