1-(tert-Butyldimethylsilyl)imidazole ≥98.0% tert-butyl-dimethyl-silylating agent

Leverantör: TCI EUROPE
Varning

Synonyms: 1-(tert-Butyldimethylsilyl)-1H-imidazole

B1043-1G B1043-5G
TCIAB1043-1GEA 605 SEK
TCIAB1043-1G TCIAB1043-5G
1-(tert-Butyldimethylsilyl)imidazole ≥98.0% tert-butyl-dimethyl-silylating agent
1-(tert-Butyldimethylsilyl)imidazole
Formel: C₉H₁₈N₂Si
MW: 182,34 g/mol
Kokpunkt: 155°C
Flampunkt: 81°C
MDL Number: MFCD00011682
CAS nummer: 54925-64-3
EINECS: 259-398-1

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (Nonaqueous Titration) min. 98.0 %
Effect test of GC derivalization Effective as derivatization agent Estradiol, Cholesterol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR