Methyl benzimidate hydrochloride ≥98.0% (by total nitrogen and titration analysis) for protein modification
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Benzimidic Acid Methyl Ester Hydrochloride
B0993-1G
B0993-5G
TCIAB0993-1GEA
510
SEK
TCIAB0993-1G
TCIAB0993-5G
Methyl benzimidate hydrochloride ≥98.0% (by total nitrogen and titration analysis) for protein modification
Methyl benzimidate hydrochloride
Formel:
C₈H₉NO MW: 171,62 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00012575 CAS nummer: 5873-90-5 EINECS: 227-537-5 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...