N,N-Bis(2-cyanoethyl)formamide ≥90.0%

Leverantör: TCI EUROPE
Fara

Synonyms: N-Formylimino-3,3'-dipropionitrile

B0977-25G B0977-5G
TCIAB0977-25GEA 2275 SEK
TCIAB0977-25G TCIAB0977-5G
N,N-Bis(2-cyanoethyl)formamide ≥90.0%
N,N-Bis(2-cyanoethyl)formamide
Formel: C₇H₉N₃O
MW: 151,17 g/mol
Kokpunkt: 122°C
MDL Number: MFCD00040894
CAS nummer: 3445-84-9
EINECS: 222-362-0
UN: 2810
ADR: 6.1,III

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR