1-Benzyl-3-p-tolyltriazene ≥98.0% for esterification
Leverantör: TCI
|
Fara
|
Synonyms:
3-p-Tolyl-1-benzyltriazene
B0949-1G
B0949-25G
TCIAB0949-1GEA
455
SEK
TCIAB0949-1G
TCIAB0949-25G
1-Benzyl-3-p-tolyltriazene ≥98.0% for esterification
1-Benzyl-3-p-tolyltriazene
Formel:
C₁₄H₁₅N₃ Storage Temperature: Kylskåp |
MDL Number:
MFCD00038226 CAS nummer: 17683-09-9 EINECS: 241-665-9 UN: 1325 ADR: 4.1,II |
Specifikation testresultat
Appearance | White to Yellow to Orange powder to crystal |
Purity (Nonaqueous Titration) | 98.0 to 102.0 % |
Melting point | 75.0 to 79.0 deg-C |
Effect test of HPLC derivatization | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...