N-Butyl-N-(4-hydroxybutyl)nitrosamine ≥90.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
1-Butanol, 4-(butylnitrosoamino)-, N-Butyl-N-nitroso-4-aminobutanol, N-Butyl-N-butan-4-ol-nitrosamine, BBN
B0938-25G
B0938-5G
TCIAB0938-25GEA
4475
SEK
TCIAB0938-25G
TCIAB0938-5G
N-Butyl-N-(4-hydroxybutyl)nitrosamine ≥90.0%
N-Butyl-N-(4-hydroxybutyl)nitrosamine
Formel:
C₈H₁₈N₂O₂ MW: 174,24 g/mol Kokpunkt: 167°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00059008 CAS nummer: 3817-11-6 |
Specifikation testresultat
Appearance | Light yellow to Amber to Dark green clear liquid |
Purity (GC) | min. 90.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...