Bis(4-chlorobutyl)ether ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 4,4'-Dichlorodibutyl Ether, 4,4'-Dichlorobutyl Ether, 4-Chlorobutyl Ether
B0929-25ML B0929-500ML
TCIAB0929-25MLEA 735 SEK
TCIAB0929-25ML TCIAB0929-500ML
Bis(4-chlorobutyl)ether ≥98.0%
Bis(4-chlorobutyl)ether
Formel: C₈H₁₆Cl₂O
MW: 199,12 g/mol
Kokpunkt: 132°C
MDL Number: MFCD00019006
CAS nummer: 6334-96-9
EINECS: 228-713-4
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %