3,3'-Diamino-N-methyldipropylamine ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: N,N-Bis(3-aminopropyl)methylamine, N-(3-Aminopropyl)-N-methyl-1,3-propanediamine
B0821-25ML B0821-500ML
TCIAB0821-25MLEA 230 SEK
TCIAB0821-25ML TCIAB0821-500ML
3,3'-Diamino-N-methyldipropylamine ≥98.0% (by GC, titration analysis)
3,3'-Diamino-N-methyldipropylamine
Formel: C₇H₁₉N₃
MW: 145,25 g/mol
Flampunkt: 105°C
MDL Number: MFCD00008217
CAS nummer: 105-83-9
EINECS: 203-336-8
UN: 2927
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be33a831cd295ec4971bff7786360be170852e71&vid=be33a831cd295ec4971bff7786360be170852e71&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %