N-Butylbenzenesulphonamide ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI EUROPE
Varning

Synonyms: N-Benzenesulphonylbutylamine

B0716-25G B0716-500G
TCIAB0716-25GEA 230 SEK
TCIAB0716-25G TCIAB0716-500G
N-Butylbenzenesulphonamide ≥98.0% (by HPLC, total nitrogen)
N-Butylbenzenesulphonamide
Formel: C₁₀H₁₅NO₂S
MW: 213,30 g/mol
Kokpunkt: 184°C
Flampunkt: 118°C
MDL Number: MFCD00025024
CAS nummer: 3622-84-2
EINECS: 222-823-6

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR