3-Butoxyphenol ≥96.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Resorcinol Monobutyl Ether
B0701-25G B0701-5G
TCIAB0701-25GEA 3200 SEK
TCIAB0701-25G TCIAB0701-5G
3-Butoxyphenol ≥96.0%
3-Butoxyphenol
Formel: C₁₀H₁₄O₂
MW: 166,22 g/mol
Kokpunkt: 269°C
MDL Number: MFCD00058976
CAS nummer: 18979-72-1
EINECS: 242-715-2
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 96.0 %