4-Bromoantipyrine ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: 4-Bromo-1,5-dimethyl-2-phenyl-1H-pyrazol-3(2H)-one

B0515-25G B0515-5G
TCIAB0515-25GEA 2575 SEK
TCIAB0515-25G TCIAB0515-5G
4-Bromoantipyrine ≥98.0% (by HPLC, titration analysis)
4-Bromoantipyrine
Formel: C₁₁H₁₁BrN₂O
MW: 267,13 g/mol
MDL Number: MFCD00020779
CAS nummer: 5426-65-3

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 115.0 to 118.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR