Copper(II) bis(2-hydroxyethyl)dithiocarbamate ≥98.0%

Leverantör: TCI

B0491-25G B0491-5G
TCIAB0491-25GEA 1675 SEK
TCIAB0491-25G TCIAB0491-5G
Copper(II) bis(2-hydroxyethyl)dithiocarbamate ≥98.0%
Copper(II) bis(2-hydroxyethyl)dithiocarbamate
Formel: C₅H₁₁NO₂S₂
MDL Number: MFCD00059149
CAS nummer: 52611-57-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bef8a8105811544eece2558bf5e024f570676ecb&vid=bef8a8105811544eece2558bf5e024f570676ecb&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Dark green to Dark blue to Black powder to crystal
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR