N-Benzylacetamide ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Lacosamide EP Impurity G, N-Acetylbenzylamine

B0404-25G B0404-500G
TCIAB0404-25GEA 1025 SEK
TCIAB0404-25G TCIAB0404-500G
N-Benzylacetamide ≥98.0%
N-Benzylacetamide
Formel: C₉H₁₁NO
MW: 149,19 g/mol
Kokpunkt: 157°C
MDL Number: MFCD00059204
CAS nummer: 588-46-5
EINECS: 209-619-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6b278d6d7285ecc4e6c5c6f08ae4411a6ce7639&vid=d6b278d6d7285ecc4e6c5c6f08ae4411a6ce7639&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 59.0 to 63.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR