2,2'-Bibenzothiazoline ≥85.0%
Leverantör: TCI
Synonyms:
2,2'-Bisbenzothiazoline, 2,2'-Dibenzothiazoline
B0377-1G
B0377-5G
TCIAB0377-1GEA
735
SEK
TCIAB0377-1G
TCIAB0377-5G
2,2'-Bibenzothiazoline ≥85.0%
2,2'-Bibenzothiazoline
Formel:
C₁₄H₁₂N₂S₂ MW: 272.38 g/mol Smältpunkt: 135°C |
MDL Number:
MFCD00059699 CAS nummer: 19258-20-9 |
Specifikation testresultat
Appearance | White to Gray to Brown powder to crystal |
Purity (Nonaqueous Titration) | min. 85.0 % |
Melting point | 140.0 to 144.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...