Benzoylthiocholine Iodide ≥98.0%

Leverantör: TCI

B0307-1G B0307-25G B0307-5G
TCIAB0307-1GEA 1125 SEK
TCIAB0307-1G TCIAB0307-25G TCIAB0307-5G
Benzoylthiocholine Iodide ≥98.0%
Benzoylthiocholine iodide
Formel: C₁₂H₁₈INOS
MDL Number: MFCD00059970
CAS nummer: 10561-14-5

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR